5-Acetylpyridin-2(1H)-one 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR903475-1G
OR903475-5G
APOSOR903475-1GEA
56.7
EUR
APOSOR903475-1G
APOSOR903475-5G
5-Acetylpyridin-2(1H)-one 97%
5-Acetylpyridin-2(1H)-one
Formel:
C₇H₇NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10697705 CAS-Nummer: 1124-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...