7-Bromoquinoline-2-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR903527-1G
OR903527-250MG
OR903527-5G
APOSOR903527-1GEA
88.6
EUR
APOSOR903527-1G
APOSOR903527-250MG
APOSOR903527-5G
7-Bromoquinoline-2-carboxylic acid 97%
7-Bromoquinoline-2-carboxylic acid
Formel:
C₁₀H₆BrNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11108693 CAS-Nummer: 1057217-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...