4,5-Dichloro-6-methylpyrimidine 95%
Lieferant: Apollo Scientific
OR903540-1G
OR903540-250MG
APOSOR903540-1GEA
140
EUR
APOSOR903540-1G
APOSOR903540-250MG
4,5-Dichloro-6-methylpyrimidine 95%
4,5-Dichloro-6-methylpyrimidine
Formel:
C₅H₄Cl₂N₂ Molecular Weight: 163,0100 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09909763 CAS-Nummer: 83942-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...