5-Brom-4-methylpyrimidin 98%
Lieferant: Apollo Scientific
OR903546-1G
OR903546-5G
APOSOR903546-1GEA
59.6
EUR
APOSOR903546-1G
APOSOR903546-5G
5-Brom-4-methylpyrimidin 98%
5-Bromo-4-methylpyrimidine
Formel:
C₅H₅BrN₂ Molecular Weight: 173,0100 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10000597 CAS-Nummer: 1439-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...