3,5-Dibrom-4-pyridinamin
Lieferant: Apollo Scientific
|
Achtung
|
OR9036-1G
OR9036-5G
APOSOR9036-1GEA
21.9
EUR
APOSOR9036-1G
APOSOR9036-5G
3,5-Dibrom-4-pyridinamin
4-Amino-3,5-dibromopyridine
Formel:
C₅H₄Br₂N₂ Molecular Weight: 251,9100 g/mol Schmelzpunkt: 165…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234048 CAS-Nummer: 84539-34-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...