4-Chlor-6-methoxychinazolin 97%
Lieferant: Apollo Scientific
OR903602-1G
OR903602-5G
APOSOR903602-1GEA
142
EUR
APOSOR903602-1G
APOSOR903602-5G
4-Chlor-6-methoxychinazolin 97%
4-Chloro-6-methoxyquinazoline
Formel:
C₉H₇ClN₂O Molecular Weight: 194,6177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657615 CAS-Nummer: 50424-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...