2-Amino-5-chloro-3-methoxypyrazine 95%
Lieferant: Apollo Scientific
OR903695-1G
OR903695-250MG
APOSOR903695-1GEA
94.4
EUR
APOSOR903695-1G
APOSOR903695-250MG
2-Amino-5-chloro-3-methoxypyrazine 95%
2-Amino-5-chloro-3-methoxypyrazine
Formel:
C₅H₆ClN₃O Molecular Weight: 159.58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11520878 CAS-Nummer: 874-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...