Thieno[2,3-b]pyrazine 97%
Lieferant: Apollo Scientific
OR903768-1G
OR903768-250MG
APOSOR903768-1GEA
502
EUR
APOSOR903768-1G
APOSOR903768-250MG
Thieno[2,3-b]pyrazine 97%
Thieno[2,3-b]pyrazine
Formel:
C₆H₄N₂S Lagertemperatur: Raumtemperatur |
CAS-Nummer:
56088-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...