5-Chlorothiophene-3-carboxylic acid 96%
Lieferant: Apollo Scientific
OR903783-5G
APOSOR903783-5GEA
163
EUR
APOSOR903783-5G
5-Chlorothiophene-3-carboxylic acid 96%
5-Chlorothiophene-3-carboxylic acid
Formel:
C5H3ClO2S Molecular Weight: 162,5900 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082877 CAS-Nummer: 36157-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...