3,5-Dichloro-4-methylaniline 98%

Lieferant: Apollo Scientific

OR903818-10G OR903818-25G OR903818-5G
APOSOR903818-10GEA 150 EUR
APOSOR903818-10G APOSOR903818-25G APOSOR903818-5G
3,5-Dichloro-4-methylaniline 98%
3,5-Dichloro-4-methylaniline
Formel: C7H7Cl2N
Molecular Weight: 176,0400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00266300
CAS-Nummer: 54730-35-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR