3-Methoxy-4-nitrophenol 96%
Lieferant: Apollo Scientific
OR903872-5G
APOSOR903872-5GEA
108
EUR
APOSOR903872-5G
3-Methoxy-4-nitrophenol 96%
3-Methoxy-4-nitrophenol
Formel:
C7H7NO4 Molecular Weight: 169,1400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11840329 CAS-Nummer: 16292-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...