Isochinolin-3-ylmethanol 98%

Lieferant: Apollo Scientific

OR903885-1G OR903885-250MG
APOSOR903885-1GEA 91.5 EUR
APOSOR903885-1G APOSOR903885-250MG
Isochinolin-3-ylmethanol 98%
Isochinolin-3-ylmethanol
Formel: C₁₀H₉NO
Molecular Weight: 159.1846 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01859798
CAS-Nummer: 76884-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR