4-(2-Chloroethyl)morpholine 98%

Lieferant: Apollo Scientific

OR903887-100G OR903887-25G
APOSOR903887-100GEA 362 EUR
APOSOR903887-100G APOSOR903887-25G
4-(2-Chloroethyl)morpholine 98%
4-(2-Chloroethyl)morpholine
Formel: C₆H₁₂ClNO
Molecular Weight: 149.6186 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00603731
CAS-Nummer: 3240-94-6
UN: 2922
ADR: 8+6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR