2-Amino-6-chlor-4-methylbenzothiazol 95%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-4-methyl-1,3-benzothiazol-2-amine

OR903958-1G OR903958-250MG
APOSOR903958-1GEA 683 EUR
APOSOR903958-1G APOSOR903958-250MG
2-Amino-6-chlor-4-methylbenzothiazol 95%
2-Amino-6-chloro-4-methylbenzothiazole
Formel: C₈H₇ClN₂S
Lagertemperatur: Raumtemperatur
CAS-Nummer: 38338-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR