3-Aminopropyldimethoxymethylsilane 95%
Lieferant: Apollo Scientific
Synonyme:
3-(Dimethoxymethylsilyl)propylamine
OR904101-100G
OR904101-500G
OR904101-5G
APOSOR904101-100GEA
183
EUR
APOSOR904101-100G
APOSOR904101-500G
APOSOR904101-5G
3-Aminopropyldimethoxymethylsilane 95%
3-Aminopropyldimethoxymethylsilane
Formel:
C₆H₁₇NO₂Si Molecular Weight: 163.29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025614 CAS-Nummer: 3663-44-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...