tert-Butyl-2-aminopropylcarbamate 97%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2-aminopropylcarbamate

OR904169-1G OR904169-5G
APOSOR904169-1GEA 159 EUR
APOSOR904169-1G APOSOR904169-5G
tert-Butyl-2-aminopropylcarbamate 97%
tert-Butyl-2-aminopropylcarbamate
Formel: C₈H₁₈N₂O₂
Molecular Weight: 174.2407 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09788609
CAS-Nummer: 255735-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR