2-(5-Methyl-2-thienyl)azepane 95%
Lieferant: Apollo Scientific
Synonyme:
2-(5-Methylthiophen-2-yl)azepane
OR904171-1G
OR904171-250MG
APOSOR904171-1GEA
1550
EUR
APOSOR904171-1G
APOSOR904171-250MG
2-(5-Methyl-2-thienyl)azepane 95%
2-(5-Methyl-2-thienyl)azepane
Formel:
C₁₁H₁₇NS Molecular Weight: 195,33 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05190319 CAS-Nummer: 527674-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...