5,7-Dimethyl-1H-indole-2-carboxylic acid 95%
Lieferant: Apollo Scientific
OR904222-100MG
OR904222-1G
OR904222-250MG
APOSOR904222-100MGEA
190
EUR
APOSOR904222-100MG
APOSOR904222-1G
APOSOR904222-250MG
5,7-Dimethyl-1H-indole-2-carboxylic acid 95%
5,7-Dimethyl-1H-indole-2-carboxylic acid
Formel:
C₁₁H₁₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02664421 CAS-Nummer: 221675-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...