2-Bromethylacetat 97%

Lieferant: Apollo Scientific

Synonyme: Essigsäure(2-bromethyl)ester

OR904265-500G
APOSOR904265-500GEA 178 EUR
APOSOR904265-500G
2-Bromethylacetat 97%
2-Bromethylacetat
Formel: C₄H₇BrO₂
Molecular Weight: 167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000235
CAS-Nummer: 927-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR