5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine
Lieferant: Apollo Scientific
OR904269-1G
OR904269-25G
OR904269-5G
APOSOR904269-1GEA
26.1
EUR
APOSOR904269-1G
APOSOR904269-25G
APOSOR904269-5G
5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine
5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine
Formel:
C₄H₇N₃OS Molecular Weight: 145,1829 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01203002 CAS-Nummer: 15884-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...