7-Nitro-3,4-dihydro-2H-1,4-benzooxazine 98%

Lieferant: Apollo Scientific

OR904351-1G OR904351-5G
APOSOR904351-1GEA 0 EUR
APOSOR904351-1G APOSOR904351-5G
7-Nitro-3,4-dihydro-2H-1,4-benzooxazine 98%
7-Nitro-3,4-dihydro-2H-1,4-benzooxazine
Formel: C₈H₈N₂O₃
Molecular Weight: 180.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11603433
CAS-Nummer: 120711-81-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR