Ethyl-5-chloro-4-methylisoxazole-3-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 5-chloro-4-methylisoxazole-3-carboxylate
OR904439-1G
OR904439-250MG
APOSOR904439-1GEA
134
EUR
APOSOR904439-1G
APOSOR904439-250MG
Ethyl-5-chloro-4-methylisoxazole-3-carboxylate 95%
Ethyl-5-chloro-4-methylisoxazole-3-carboxylate
Formel:
C₇H₈ClNO₃ Molecular Weight: 189,6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11520431 CAS-Nummer: 3356-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...