2-Methylpyrimidine-4-carbaldehyde 98%

Lieferant: Apollo Scientific
Gefahr

OR904478-1G OR904478-250MG OR904478-5G
APOSOR904478-1GEA 194 EUR
APOSOR904478-1G APOSOR904478-250MG APOSOR904478-5G
2-Methylpyrimidine-4-carbaldehyde 98%
2-Methylpyrimidine-4-carbaldehyde
Formel: C₆H₆N₂O
Molecular Weight: 122,13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09832936
CAS-Nummer: 1004-17-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR