Ethyl-2-oxobutanoate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-oxobutanoate, Ethyl 2-oxobutyrate

OR904493-100G OR904493-25G
APOSOR904493-100GEA 405 EUR
APOSOR904493-100G APOSOR904493-25G
Ethyl-2-oxobutanoate 95%
Ethyl-2-oxobutanoate
Formel: C₆H₁₀O₃
Molecular Weight: 130,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03265336
CAS-Nummer: 15933-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR