1H-Pyrrol-1-ylacetic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-(1H-Pyrrol-1-yl)acetic acid

OR904643-1G OR904643-250MG
APOSOR904643-1GEA 279 EUR
APOSOR904643-1G APOSOR904643-250MG
1H-Pyrrol-1-ylacetic acid 98%
1H-Pyrrol-1-ylacetic acid
Formel: C₆H₇NO₂
Molecular Weight: 125,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00955795
CAS-Nummer: 19167-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR