Methyl-3-hydroxy-5-methyl-2-thiophenecarboxylate 95%
Lieferant: Apollo Scientific
OR904800-1G
OR904800-250MG
APOSOR904800-1GEA
841
EUR
APOSOR904800-1G
APOSOR904800-250MG
Methyl-3-hydroxy-5-methyl-2-thiophenecarboxylate 95%
Methyl-3-hydroxy-5-methyl-2-thiophenecarboxylate
Formel:
C₇H₈O₃S Lagertemperatur: Raumtemperatur |
CAS-Nummer:
5556-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...