4-Methoxybenzylisocyanid 95%
Lieferant: Apollo Scientific
Synonyme:
1-(Isocyanomethyl)-4-methoxybenzene
OR904817-1G
OR904817-250MG
APOSOR904817-1GEA
955
EUR
APOSOR904817-1G
APOSOR904817-250MG
4-Methoxybenzylisocyanid 95%
4-Methoxybenzylisocyanid
Formel:
C₉H₉NO Molecular Weight: 147.173 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02664572 CAS-Nummer: 1197-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...