Methyl-2-bromoisovalerate 98%

Lieferant: Apollo Scientific

OR904870-10G OR904870-1G OR904870-5G
APOSOR904870-10GEA 451 EUR
APOSOR904870-10G APOSOR904870-1G APOSOR904870-5G
Methyl-2-bromoisovalerate 98%
Methyl-2-bromoisovalerate
Formel: C₆H₁₁BrO₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 26330-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR