6-Methylpyridine-3-sulphonic acid 98%

Lieferant: Apollo Scientific

OR904880-1G OR904880-5G
APOSOR904880-1GEA 59.6 EUR
APOSOR904880-1G APOSOR904880-5G
6-Methylpyridine-3-sulphonic acid 98%
6-Methylpyridine-3-sulphonic acid
Formel: C₆H₇NO₃S
Molecular Weight: 173,1897 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01646153
CAS-Nummer: 4808-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR