4-Brom-5-chlor-2-methylanilin 97%

Lieferant: Apollo Scientific
Achtung

OR904891-100G OR904891-25G
APOSOR904891-100GEA 95.7 EUR
APOSOR904891-100G APOSOR904891-25G
4-Brom-5-chlor-2-methylanilin 97%
4-Brom-5-chlor-2-methylanilin
Formel: C₇H₇BrClN
Molecular Weight: 220,5 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07782182
CAS-Nummer: 30273-47-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR