3-Amino-6-chloro-2-pyrazinecarbonitrile 95%
Lieferant: Apollo Scientific
OR904953-100MG
OR904953-1G
OR904953-250MG
APOSOR904953-100MGEA
56.7
EUR
APOSOR904953-100MG
APOSOR904953-1G
APOSOR904953-250MG
3-Amino-6-chloro-2-pyrazinecarbonitrile 95%
3-Amino-6-chloro-2-pyrazinecarbonitrile
Formel:
C5H3ClN4 Molecular Weight: 154,5600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10697784 CAS-Nummer: 17231-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...