Methyl-6-cyanpicolinat 95%
Lieferant: Apollo Scientific
Synonyme:
6-Cyano-2-pyridine carboxylic acid methyl ester, Methyl 6-cyanopicolinate
OR905007-1G
OR905007-250MG
APOSOR905007-1GEA
68.3
EUR
APOSOR905007-1G
APOSOR905007-250MG
Methyl-6-cyanpicolinat 95%
Methyl-6-cyanpicolinat
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,1500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07367924 CAS-Nummer: 98436-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...