2-Bromisobuttersäure 95%
Lieferant: Apollo Scientific
Synonyme:
2-Brom-2-methylpropionsäure, 2-Brom-2-methylpropansäure, α-Bromisobuttersäure
OR905022-100G
OR905022-1KG
OR905022-500G
APOSOR905022-100GEA
132
EUR
APOSOR905022-100G
APOSOR905022-1KG
APOSOR905022-500G
2-Bromisobuttersäure 95%
2-Bromisobuttersäure
Formel:
C₄H₇BrO₂ Molecular Weight: 167 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004168 CAS-Nummer: 2052-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...