2-Chlorothiazolo[4,5-c]pyridine 97%

Lieferant: Apollo Scientific
Achtung

OR905191-1G OR905191-250MG
APOSOR905191-1GEA 265 EUR
APOSOR905191-1G APOSOR905191-250MG
2-Chlorothiazolo[4,5-c]pyridine 97%
2-Chlorothiazolo[4,5-c]pyridine
Formel: C₆H₃ClN₂S
Lagertemperatur: Kühlschrank
MDL: MFCD13193436
CAS-Nummer: 884860-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR