3-(1-Methyl-1H-pyrazol-4-yl)propionic acid 97%

Lieferant: Apollo Scientific

OR905214-1G OR905214-250MG OR905214-5G
APOSOR905214-1GEA 255 EUR
APOSOR905214-1G APOSOR905214-250MG APOSOR905214-5G
3-(1-Methyl-1H-pyrazol-4-yl)propionic acid 97%
3-(1-Methyl-1H-pyrazol-4-yl)propionic acid
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04969921
CAS-Nummer: 796845-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR