4-Aminomethylquinoline hydrochloride 95%

Lieferant: Apollo Scientific

OR905248-1G OR905248-250MG
APOSOR905248-1GEA 236 EUR
APOSOR905248-1G APOSOR905248-250MG
4-Aminomethylquinoline hydrochloride 95%
4-Aminomethylquinoline hydrochloride
Formel: C₁₀H₁₁ClN₂
Molecular Weight: 194.6600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05861486
CAS-Nummer: 1095661-17-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR