2-(3-Methylphenyl)pyrrolidine 95%

Lieferant: Apollo Scientific

OR905257-1G OR905257-5G
APOSOR905257-1GEA 50.9 EUR
APOSOR905257-1G APOSOR905257-5G
2-(3-Methylphenyl)pyrrolidine 95%
2-(3-Methylphenyl)pyrrolidine
Formel: C₁₁H₁₅N
Molecular Weight: 161.2400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02663444
CAS-Nummer: 72216-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR