1-(2-Ethylphenyl)-1H-pyrrole 95%
Lieferant: Apollo Scientific
OR905271-1G
OR905271-250MG
APOSOR905271-1GEA
458
EUR
APOSOR905271-1G
APOSOR905271-250MG
1-(2-Ethylphenyl)-1H-pyrrole 95%
1-(2-Ethylphenyl)-1H-pyrrole
Formel:
C₁₂H₁₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD02665229 CAS-Nummer: 299426-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...