Piroctone olamine

Lieferant: Apollo Scientific

Synonyme: 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine, 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2(1H)-one 2-aminoethanol salt

OR905511-25G OR905511-5G OR905511-100G
APOSOR905511-25GEA 172 EUR
APOSOR905511-25G APOSOR905511-5G APOSOR905511-100G
Piroctone olamine
Piroctone olamine
Formel: C₁₄H₂₃NO₂
Molecular Weight: 298,43 g/mol
Siedepunkt: 130…135 °C
Dichte: 1,1 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD01690792
CAS-Nummer: 68890-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR