5-(Bromomethyl)-1-methyl-1H-benzo[d][1,2,3]triazole 95+%

Lieferant: Apollo Scientific
Achtung

OR905560-1G OR905560-250MG OR905560-5G
APOSOR905560-1GEA 596 EUR
APOSOR905560-1G APOSOR905560-250MG APOSOR905560-5G
5-(Bromomethyl)-1-methyl-1H-benzo[d][1,2,3]triazole 95+%
5-(Bromomethyl)-1-methyl-1H-benzo[d][1,2,3]triazole
Formel: C₈H₈BrN₃
Schmelzpunkt: 120 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 499770-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR