2-Amino-3-methoxybenzaldehyde 95%

Lieferant: Apollo Scientific

OR905592-1G OR905592-250MG OR905592-5G
APOSOR905592-1GEA 178 EUR
APOSOR905592-1G APOSOR905592-250MG APOSOR905592-5G
2-Amino-3-methoxybenzaldehyde 95%
2-Amino-3-methoxybenzaldehyde
Formel: C₈H₉NO₂
Molecular Weight: 151,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09999245
CAS-Nummer: 70127-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR