Ethyl-2-ethylacetoacetat 98%

Lieferant: Apollo Scientific

Synonyme: 2-Ethyl-3-oxobuttersäureethylester, 2-Ethylacetessigsäureethylester

OR905609-100G OR905609-500G
APOSOR905609-100GEA 92.8 EUR
APOSOR905609-100G APOSOR905609-500G
Ethyl-2-ethylacetoacetat 98%
Ethyl-2-ethylacetoacetat
Formel: C₈H₁₄O₃
Molecular Weight: 158.2 g/mol
Dichte: 0.9937
Lagertemperatur: Raumtemperatur
MDL: MFCD00039898
CAS-Nummer: 607-97-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR