1-Cyclobutylethanol 95%
Lieferant: Apollo Scientific
OR905673-1G
OR905673-250MG
OR905673-5G
APOSOR905673-1GEA
144
EUR
APOSOR905673-1G
APOSOR905673-250MG
APOSOR905673-5G
1-Cyclobutylethanol 95%
1-Cyclobutylethanol
Formel:
C₆H₁₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061039 CAS-Nummer: 7515-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...