2-Bromo-4,5-dimethylthiazole 98%

Lieferant: Apollo Scientific

OR905682-5G
APOSOR905682-5GEA 59.6 EUR
APOSOR905682-5G
2-Bromo-4,5-dimethylthiazole 98%
2-Bromo-4,5-dimethylthiazole
Formel: C₅H₆BrNS
Molecular Weight: 192,0768 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501629
CAS-Nummer: 29947-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR