1-Boc-3-piperidon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-BOC-3-piperidone, 1-Boc-3-Piperidinone
OR9057-100G
OR9057-25G
OR9057-5G
APOSOR9057-100GEA
75.4
EUR
APOSOR9057-100G
APOSOR9057-25G
APOSOR9057-5G
1-Boc-3-piperidon 98%
1-Boc-3-piperidon
Formel:
C₁₀H₁₇NO₃ Molecular Weight: 199,25 g/mol Schmelzpunkt: 35…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631193 CAS-Nummer: 98977-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...