5-Amino-2-bromopyrimidine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR905856-1G
OR905856-250MG
APOSOR905856-1GEA
84.1
EUR
APOSOR905856-1G
APOSOR905856-250MG
5-Amino-2-bromopyrimidine 95%
5-Amino-2-bromopyrimidine
Formel:
C₄H₄BrN₃ Molecular Weight: 173,9987 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07778608 CAS-Nummer: 56621-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...