2-Thiazoleacetic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Thiazolyl)acetic Acid

OR905873-1G OR905873-250MG
APOSOR905873-1GEA 589 EUR
APOSOR905873-1G APOSOR905873-250MG
2-Thiazoleacetic acid 95%
2-Thiazoleacetic acid
Formel: C₅H₅NO₂S
Molecular Weight: 143,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02094168
CAS-Nummer: 188937-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR