5-Pyrazol-1-ylmethylfuran-2-carboxylic acid 95%
Lieferant: Apollo Scientific
Synonyme:
5-(1H-Pyrazol-1-ylmethyl)-2-furoic acid
OR905916-1G
OR905916-5G
APOSOR905916-1GEA
219
EUR
APOSOR905916-1G
APOSOR905916-5G
5-Pyrazol-1-ylmethylfuran-2-carboxylic acid 95%
5-Pyrazol-1-ylmethylfuran-2-carboxylic acid
Formel:
C₉H₈N₂O₃ Molecular Weight: 192,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02253715 CAS-Nummer: 386736-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...