N-Methyl-p-toluidin 98%
Lieferant: Apollo Scientific
Synonyme:
1-Methyl-4-(methylamino)benzol, N,4-Dimethylanilin, 4-Methyl-N-methylanilin, N-Methyl-4-methylanilin, p-(Methylamino)toluol, 4-(Methylamino)toluol
OR905934-100G
OR905934-25G
APOSOR905934-100GEA
43.5
EUR
APOSOR905934-100G
APOSOR905934-25G
N-Methyl-p-toluidin 98%
N-Methyl-p-toluidin
Formel:
C₈H₁₁N Molecular Weight: 121.18 g/mol Siedepunkt: 89 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025628 CAS-Nummer: 623-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...