4-(Pyridin-3-yloxy)aniline 95%

Lieferant: Apollo Scientific

Synonyme: 4-(Pyridin-3-yloxy)-phenylamine

OR905968-1G OR905968-250MG
APOSOR905968-1GEA 82.8 EUR
APOSOR905968-1G APOSOR905968-250MG
4-(Pyridin-3-yloxy)aniline 95%
4-(Pyridin-3-yloxy)aniline
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186.2100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07186369
CAS-Nummer: 80650-45-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR